Iron(III) phosphate hydrate
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Iron phosphate hydrate, Iron (III) orthophosphate hydrate
93-2647-500G
93-2647-2000G
100209-422EA
147.2
USD
100209-422
100199-536
Iron(III) phosphate hydrate
Iron(III) phosphate hydrate
Formula:
FeO₄P·xH₂O MW: 150.82 g/mol (anhy) |
MDL Number:
MFCD00016096 CAS Number: 51833-68-2 EINECS: 233-149-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...