Iron(II) bromide, anhydrous ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Iron dibromide, Ferrous bromide, Iron(II) bromide

93-2629-10G 93-2629-50G
100204-688EA 116.06 USD
100204-688 100200-086
Iron(II) bromide, anhydrous ≥98%
Iron(II) bromide
Formula: Br₂Fe
MW: 215.66 g/mol
Melting Pt: 684 °C (dec.)
Density: 4.636
MDL Number: MFCD00016081
CAS Number: 7789-46-0
EINECS: 232-168-8
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR