Iron(II,III) oxide, black ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

93-2616-500G 93-2616-2000G
100202-020EA 66.52 USD
100202-020 100201-086
Iron(II,III) oxide, black ≥95%
Iron(II,III) oxide
Formula: Fe₃O₄
MW: 231.54 g/mol
Melting Pt: 1560 °C
Density: 5.18 g/cm³ (20 °C)
MDL Number: MFCD00011010
CAS Number: 1317-61-9
EINECS: 215-277-5
Merck Index: 12,04087

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR