Iron(III) nitrate nonahydrate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate
93-2614-5000G
93-2614-250G
93-2614-1000G
100207-808EA
172.68
USD
100207-808
100205-456
100200-084
Iron(III) nitrate nonahydrate ≥98%
Iron(III) nitrate nonahydrate
Formula:
Fe(NO₃)₃·9H₂O MW: 404 g/mol Boiling Pt: 125 (dec.) Melting Pt: 47 Density: 1.684 Storage Temperature: Ambient |
MDL Number:
MFCD00149708 CAS Number: 7782-61-8 EINECS: 233-899-5 UN: 1466 ADR: 5.1 III Merck Index: 13,04057 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...