Iron(III) fluoride, anhydrous ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Iron trifluoride, Ferric fluoride

93-2610-50G 93-2610-10G
100203-422EA 169.85 USD
100203-422 100210-288
Iron(III) fluoride, anhydrous ≥99%
Iron(III) fluoride
Formula: F₃Fe
MW: 112.84 g/mol
Melting Pt: > 1000° (subl.)
Density: 3.52 g/cm³ (20 °C)
MDL Number: MFCD00011012
CAS Number: 7783-50-8
EINECS: 232-002-4
UN: 1759
ADR: 8,III
Merck Index: 12,04065

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR