Manganese(III) fluoride ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-2542-25G
93-2542-5G
100209-052EA
175.51
USD
100209-052
100207-802
Manganese(III) fluoride ≥98%
Manganese(III) fluoride
Formula:
F₃Mn MW: 111.93 g/mol Melting Pt: Dec. Density: 3.54 g/cm³ (20 °C) |
MDL Number:
MFCD00016223 CAS Number: 7783-53-1 EINECS: 232-006-6 UN: 3288 ADR: 6.1,III Merck Index: 12,05784 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...