Manganese(II) chloride, anhydrous ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Manganese dichloride
93-2527-2000G
93-2527-500G
100200-082EA
113.23
USD
100200-082
100204-254
Manganese(II) chloride, anhydrous ≥97%
Manganese(II) chloride
Formula:
MnCl₂ MW: 125.84 g/mol Boiling Pt: 1190 °C (1013 hPa) Melting Pt: 650 °C Density: 2.98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011114 CAS Number: 7773-01-5 EINECS: 231-869-6 UN: 3288 ADR: 6.1,III Merck Index: 12,05768 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...