Manganese(III) oxide ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Manganese oxide (Mn2O3), Dimanganese trioxide

93-2522-1000G 93-2522-250G
100200-542EA 390.65 USD
100200-542 100206-980
Manganese(III) oxide ≥99%
Manganese(III) oxide
Formula: Mn₂O₃
MW: 157.87
Melting Pt: 1347 (dec.)
Density: 4.50
MDL Number: MFCD00016217
CAS Number: 1317-34-6
EINECS: 215-264-4
Merck Index: 13,05761

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR