Manganese(II) oxide ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Manganese(ii)oxide, Manganese oxide

93-2515-2000G 93-2515-500G
100209-416EA 234.95 USD
100209-416 100202-014
Manganese(II) oxide ≥99%
Manganese(II) oxide
Formula: MnO
MW: 70.94 g/mol
Melting Pt: 1840 °C
Density: 5.43 g/cm³ (20 °C)
MDL Number: MFCD00016226
CAS Number: 1344-43-0
EINECS: 215-695-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR