Manganese(II,III) oxide ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-2513-1000G
93-2513-250G
100199-532EA
437.35
USD
100199-532
100196-566
Manganese(II,III) oxide ≥97%
Manganese(II,III) oxide
Formula:
Mn₃O₄ MW: 228.81 Melting Pt: 1705 °C Density: 4.856 |
MDL Number:
MFCD00016218 CAS Number: 1317-35-7 EINECS: 215-266-5 Merck Index: 13,05753 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...