Manganese(II) iodide, anhydrous ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Manganous iodide

93-2512-50G 93-2512-10G
100207-280EA 717.6 USD
100207-280 100211-232
Manganese(II) iodide, anhydrous ≥98%
Manganese(II) iodide
Formula: I₂Mn
MW: 308.75 g/mol
Melting Pt: 638
Density: 5.04 g/cm³ (20 °C)
MDL Number: MFCD00016225
CAS Number: 7790-33-2
EINECS: 232-201-6
Merck Index: 12,05772

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR