Manganese(IV) oxide ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid

93-2510-100G 93-2510-500G
100200-540EA 94.83 USD
100200-540 100210-682
Manganese(IV) oxide ≥99%
Manganese(IV) oxide
Formula: MnO₂
MW: 86.94 g/mol
Melting Pt: 535 (dec.)
Density: 5.026
MDL Number: MFCD00003463
CAS Number: 1313-13-9
EINECS: 215-202-6
UN: 1479
ADR: 5.1 II
Merck Index: 14,05730

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR