Manganese(II) fluoride ≥99%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Manganese(II) difluoride, Manganese difluoride
93-2509-25G
93-2509-5G
100202-012EA
100.49
USD
100202-012
100197-768
Manganese(II) fluoride ≥99%
Manganese(II) fluoride
Formula:
F₂Mn MW: 92.93 Melting Pt: 856 °C Density: 3.98 |
MDL Number:
MFCD00016222 CAS Number: 7782-64-1 EINECS: 231-960-0 UN: 3288 ADR: 6.1 III Merck Index: 12,05769 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...