Manganese(II) bromide tetrahydrate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Manganous bromide tetrahydrate, Manganese dibromide tetrahydrate, Manganese(II) bromide hydrate
93-2507-25G
93-2507-100G
100211-230EA
127.38
USD
100211-230
100200-328
Manganese(II) bromide tetrahydrate ≥98%
Manganese(II) bromide tetrahydrate
Formula:
Br₂Mn·4H₂O MW: 286.81 g/mol Melting Pt: 64.3 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149791 CAS Number: 10031-20-6 EINECS: 236-591-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...