Chromium ≥99%, powder

Supplier: STREM CHEMICALS INC MS
Warning

93-2458-500G 93-2458-100G
100202-008EA 191.08 USD
100202-008 100196-564
Chromium ≥99%, powder
Chromium
Formula: Cr
MW: 52
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.20
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
UN: 3089
ADR: 4.1 III
Merck Index: 12,02288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR