Chromium ≥99.8%, chips
Supplier: STREM CHEMICALS INC MS
93-2457-250G
93-2457-50G
100199-526EA
161.35
USD
100199-526
100208-532
Chromium ≥99.8%, chips
Chromium
Formula:
Cr MW: 52 Boiling Pt: 2469 Melting Pt: 1840 Density: 7.20 Storage Temperature: Ambient |
MDL Number:
MFCD00010944 CAS Number: 7440-47-3 EINECS: 231-157-5 Merck Index: 12,02288 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...