Chromium(III) oxide ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Chromic oxide

93-2438-250G 93-2438-50G
100199-524EA 159.94 USD
100199-524 100205-440
Chromium(III) oxide ≥99%
Chromium(III) oxide
Formula: Cr₂O₃
MW: 151.99 g/mol
Boiling Pt: 4000
Melting Pt: 2435
Density: 5.21
Storage Temperature: Ambient
MDL Number: MFCD00010949
CAS Number: 1308-38-9
EINECS: 215-160-9
Merck Index: 13,02247

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR