Chromium(III) oxide ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Chromic oxide

93-2437-1000G 93-2437-250G
100199-522EA 141.54 USD
100199-522 100199-020
Chromium(III) oxide ≥98%
Chromium(III) oxide
Formula: Cr₂O₃
MW: 151.99 g/mol
Boiling Pt: 4000 °C
Melting Pt: 2435 °C
Density: 5.22 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010949
CAS Number: 1308-38-9
EINECS: 215-160-9
Merck Index: 13,02247

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR