Chromium(III) fluoride, anhydrous ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Chromium trifluoride

93-2434-25G 93-2434-1G 93-2434-5G
100208-142EA 992.18 USD
100208-142 100197-762 100196-558
Chromium(III) fluoride, anhydrous ≥98%
Chromium(III) fluoride
Formula: CrF₃
MW: 108.99
Melting Pt: 1000
Density: 3.8 g/cm³ (20 °C)
MDL Number: MFCD00016045
CAS Number: 7788-97-8
EINECS: 232-137-9
UN: 1756
ADR: 8 II
Merck Index: 12,02279

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR