Chromium(II) fluoride, anhydrous ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

93-2433-2G 93-2433-10G
100210-274EA 128.8 USD
100210-274 100199-018
Chromium(II) fluoride, anhydrous ≥95%
Chromium(II) fluoride
Formula: CrF₂
MW: 89.99 g/mol
Boiling Pt: 1300 °C (1013 hPa)
Melting Pt: 1100 °C
Density: 4.11 g/cm³ (20 °C)
MDL Number: MFCD00016044
CAS Number: 10049-10-2
EINECS: 233-168-0
UN: 1759
ADR: 8,II
Merck Index: 12,02302

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR