Chromium(II) chloride, anhydrous ≥99.9% (Cr basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Chromium dichloride
93-2405-25G
93-2405-5G
100200-076EA
468.49
USD
100200-076
100203-418
Chromium(II) chloride, anhydrous ≥99.9% (Cr basis)
Chromium(II) chloride
Formula:
Cl₂Cr MW: 122.9 g/mol Boiling Pt: 1302 °C (1013 hPa) Melting Pt: 824 °C Density: 2.878 Storage Temperature: Ambient |
MDL Number:
MFCD00010947 CAS Number: 10049-05-5 EINECS: 233-163-3 UN: 3260 ADR: 8 III Merck Index: 13,02264 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...