Vanadium(V) oxide ≥99.99% (V basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide

93-2337-5G 93-2337-25G
100204-684EA 127.38 USD
100204-684 100206-144
Vanadium(V) oxide ≥99.99% (V basis), PURATREM
Vanadium(V) oxide
Formula: O₅V₂
MW: 181.88 g/mol
Boiling Pt: 1750 °C (1013 hPa)
Melting Pt: 690 °C
Density: 3.35 g/cm³ (20 °C)
MDL Number: MFCD00011457
CAS Number: 1314-62-1
EINECS: 215-239-8
UN: 2862
ADR: 6.1,III
Merck Index: 13,09987

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR