Vanadium(III) bromide ≥99.7% (V basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-2335-25G
93-2335-1G
93-2335-5G
100198-336EA
1409.72
USD
100198-336
100199-016
100202-442
Vanadium(III) bromide ≥99.7% (V basis)
Vanadium(III) bromide
Formula:
Br₃V MW: 290.65 g/mol Density: 4 g/cm³ (20 °C) |
MDL Number:
MFCD00049695 CAS Number: 13470-26-3 EINECS: 236-736-6 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...