Vanadyl sulfate hydrate

Supplier: STREM CHEMICALS INC MS
Danger

93-2334-25G 93-2334-100G
100200-074EA 84.92 USD
100200-074 100196-550
Vanadyl sulfate hydrate
Vanadyl sulphate hydrate
Formula: O₅SV·H₂O
MDL Number: MFCD00149933
CAS Number: 123334-20-3
EINECS: 248-652-7
UN: 2931
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR