Vanadium oxytrifluoride ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Vanadium (V) oxide fluoride, Vanadium (V) trifluoride oxide, Vanadium oxyfluoride, Vanadyl trifluoride, Vanadium trifluoride oxide, Trifluorooxovanadium

93-2332-25G 93-2332-5G
100203-416EA 292.98 USD
100203-416 100209-050
Vanadium oxytrifluoride ≥98%
Vanadium oxytrifluoride
Formula: F₃OV
MW: 123.94 g/mol
Boiling Pt: 480 °C
Melting Pt: 300 °C
Density: 2.459 g/cm³ (21 °C)
MDL Number: MFCD00042555
CAS Number: 13709-31-4
EINECS: 237-250-7
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR