Ammonium metavanadate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Ammonium monovanadate, Ammonium vanadium trioxide, Ammonium vanadium oxide

93-2301-500G 93-2301-100G
100201-080EA 263.26 USD
100201-080 100197-756
Ammonium metavanadate ≥99%
Ammonium metavanadate
Formula: NH₄VO₃
MW: 116.98 g/mol
Melting Pt: 200 (dec.)
Density: 2.33
MDL Number: MFCD00011430
CAS Number: 7803-55-6
EINECS: 232-261-3
UN: 2859
ADR: 6.1 II
Merck Index: 13,00572

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR