Titanium ≥99%, powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-2267-250G
93-2267-50G
100202-534EA
175.51
USD
100202-534
100206-972
Titanium ≥99%, powder
Titanium
Formula:
Ti Boiling Pt: 3277 °C (1013 hPa) Melting Pt: 1677 °C Density: 4.5 g/cm³ (20 °C) |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 UN: 2546 ADR: 4.2,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...