Titanium ≥99.97%, wire

Supplier: STREM CHEMICALS INC MS
Warning

93-2256-100CM 93-2256-25CM
100205-870EA 247.69 USD
100205-870 100205-422
Titanium ≥99.97%, wire
Titanium
Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR