Titanium ≥99.99%, wire
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-2255-50CM
93-2255-250CM
100201-986EA
147.2
USD
100201-986
100199-274
Titanium ≥99.99%, wire
Titanium
Formula:
Ti Boiling Pt: 3262 °C (1013 hPa) Melting Pt: 1660 °C Density: 4.51 g/cm³ (20 °C) |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 Merck Index: 12,09610 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...