Titanyl(IV) acetylacetonate ≥95%

Supplier: STREM CHEMICALS INC MS

Synonyms: Titanium(IV) oxide acetylacetonate, Titanium(IV) oxyacetylacetonate, Titanyl Acetylacetonate, Titanium (IV) oxide bis(acetylacetonate), Oxobis(2,4-pentanedionato-O,O')titanium (IV), Titanium (IV) oxide bis(2,4-pentanedionate)

93-2225-100G 93-2225-5G 93-2225-25G
100200-070EA 607.2 USD
100200-070 100204-234 100211-228
Titanyl(IV) acetylacetonate ≥95%
Titanyl(IV) acetylacetonate
Formula: C₁₀H₁₄O₅Ti
MW: 262.1 g/mol
Melting Pt: 184 °C (dec.)
MDL Number: MFCD00013505
CAS Number: 14024-64-7
EINECS: 237-861-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR