Titanium(IV) fluoride ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-2222-50G
93-2222-10G
100198-332EA
206.65
USD
100198-332
100197-746
Titanium(IV) fluoride ≥98%
Titanium(IV) fluoride
Halides for CVD & ALD
Formula:
F₄Ti Boiling Pt: 284 °C (1013 hPa) Melting Pt: 400 °C Density: 2.798 g/cm³ (20.5 °C) |
MDL Number:
MFCD00042545 CAS Number: 7783-63-3 EINECS: 232-017-6 UN: 1759 ADR: 8,II Merck Index: 12,09619 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...