Titanium nitride ≥99% (Ti basis)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Titanium nitride
93-2213-100G
93-2213-25G
100208-134EA
209.48
USD
100208-134
100205-416
Titanium nitride ≥99% (Ti basis)
Titanium nitride
Formula:
NTi MW: 61.89 g/mol Melting Pt: 2930 °C Density: 5.24 g/cm³ (25 °C) |
MDL Number:
MFCD00049596 CAS Number: 25583-20-4 EINECS: 247-117-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...