Titanium(II) oxide ≥99.9% (Ti basis), pellets, Optical Grade
Supplier: STREM CHEMICALS INC MS
Synonyms:
Titanium oxide, Titanium monoxide
93-2212-25G
93-2212-5G
93-2212-100G
100199-512EA
157.11
USD
100199-512
100205-414
100210-258
Titanium(II) oxide ≥99.9% (Ti basis), pellets, Optical Grade
Titanium(II) oxide
Formula:
OTi MW: 63.88 g/mol Boiling Pt: 3660 °C (1013 hPa) Melting Pt: 1750 °C Density: 4.88 g/cm³ (20 °C) |
MDL Number:
MFCD00036279 CAS Number: 12137-20-1 EINECS: 235-236-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...