Titanium(IV) ethanolate (contains 5-15% isopropanol)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Titanium (IV) ethoxide, Titanium tetraethanolate, Tetraethyl orthotitanate

93-2209-100G 93-2209-25G 93-2209-500G
100210-256EA 113.23 USD
100210-256 100205-412 100205-868
Titanium(IV) ethanolate (contains 5-15% isopropanol)
Titanium(IV) ethanolate

Alkoxide

Formula: C₈H₂₀O₄Ti
MW: 228.12 g/mol
Boiling Pt: 150 °C (1013 hPa)
Density: 1.107 g/cm³ (20 °C)
Flash Pt: 33 °C
MDL Number: MFCD00009071
CAS Number: 3087-36-3
EINECS: 221-410-8
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR