Titanium(IV) oxide ≥99.5%, sintered lumps
Supplier: STREM CHEMICALS INC MS
Synonyms:
Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide
93-2208-100G
93-2208-25G
100199-008EA
482.65
USD
100199-008
100210-254
Titanium(IV) oxide ≥99.5%, sintered lumps
Titanium(IV) oxide
Formula:
TiO₂ MW: 79.88 g/mol Boiling Pt: 2900 °C (1013 hPa) Melting Pt: 1855 °C Density: 4.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011269 CAS Number: 13463-67-7 EINECS: 236-675-5 Merck Index: 13,09549 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...