Titanium carbide ≥99%

Supplier: STREM CHEMICALS INC MS

93-2205-100G 93-2205-500G
100208-518EA 174.09 USD
100208-518 100203-410
Titanium carbide ≥99%
Titanium carbide
Formula: CTi
MW: 59.89 g/mol
Boiling Pt: 4300
Melting Pt: 3067
Density: 4.93
MDL Number: MFCD00011268
CAS Number: 12070-08-5
EINECS: 235-120-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR