Ammonium hexafluorotitanate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Ammonium fluotitanate, Ammonium hexafluorotitanate, Ammonium hexafluorotitanate(IV), Ammonium hexafluorotitanate (IV)
93-2202-100G
93-2202-25G
100199-272EA
522.28
USD
100199-272
100207-796
Ammonium hexafluorotitanate ≥98%
Ammonium hexafluorotitanate
Formula:
H₈F₆N₂Ti MW: 197.95 g/mol |
MDL Number:
MFCD00010888 CAS Number: 16962-40-6 EINECS: 241-036-9 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...