Scandium(III) fluoride ≥99.9% (Sc basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-2112-5G
93-2112-1G
100196-890EA
970.95
USD
100196-890
100208-132
Scandium(III) fluoride ≥99.9% (Sc basis)
Scandium(III) fluoride
Formula:
F₃Sc MW: 101.95 g/mol Boiling Pt: 1527 °C (1013 hPa) Melting Pt: 1140 - 1160 °C Density: 2.52 g/cm³ (25 °C) |
MDL Number:
MFCD00016325 CAS Number: 13709-47-2 EINECS: 237-255-4 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...