Scandium(III) chloride hexahydrate ≥99.9% (Sc basis)
Supplier: STREM CHEMICALS INC MS
93-2111-5G
93-2111-1G
100206-960EA
485.48
USD
100206-960
100206-958
Scandium(III) chloride hexahydrate ≥99.9% (Sc basis)
Scandium(III) chloride hexahydrate
Formula:
Cl₃Sc·6H₂O MW: 259.41 g/mol |
MDL Number:
MFCD00149852 CAS Number: 20662-14-0 EINECS: 233-799-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...