Scandium(III) nitrate hydate ≥99.9% (Sc basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Trinitric acid scandium salt hydrate, Scandium(3+) trinitrate hydrate, Scandium trinitrate hydrate
93-2104-5G
93-2104-1G
100207-462EA
730.34
USD
100207-462
100209-044
Scandium(III) nitrate hydate ≥99.9% (Sc basis)
Scandium(III) nitrate hydrate
Formula:
Sc(NO₃)₃·xH₂O MW: 230.97 g/mol (anhy) |
MDL Number:
MFCD00011220 CAS Number: 13465-60-6 EINECS: 236-701-5 UN: 1477 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...