Scandium(III) chloride, anhydrous ≥99.99% (Sc basis), PURATREM, sublimated
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-2102-1G
93-2102-5G
100197-736EA
213.72
USD
100197-736
100209-042
Scandium(III) chloride, anhydrous ≥99.99% (Sc basis), PURATREM, sublimated
Scandium(III) chloride
Formula:
Cl₃Sc MW: 151.31 g/mol Melting Pt: 939 °C Density: 1.67 g/cm³ (940 °C) |
MDL Number:
MFCD00011221 CAS Number: 10361-84-9 EINECS: 233-799-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...