Calcium ≥99.5%, granules
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-2075-100G
93-2075-500G
100210-248EA
107.57
USD
100210-248
100203-844
Calcium ≥99.5%, granules
Calcium
Formula:
Ca Boiling Pt: 1484 °C (1013 hPa) Melting Pt: 850 °C Density: 1.54 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085314 CAS Number: 7440-70-2 EINECS: 231-179-5 UN: 1401 ADR: 4.3,II Merck Index: 13,01644 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...