Calcium sulfide ≥99.9% (Ca basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-2047-10G
93-2047-50G
100200-736EA
116.06
USD
100200-736
100200-322
Calcium sulfide ≥99.9% (Ca basis)
Calcium sulphide
Formula:
CaS MW: 72.14 g/mol Melting Pt: ? 1000 °C Density: 2.59 g/cm³ (20 °C) |
MDL Number:
MFCD00015990 CAS Number: 20548-54-3 EINECS: 243-873-5 UN: 3132 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...