Calcium stearate ≥85%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Stearic acid calcium salt
93-2045-100G
93-2045-1000G
100201-964EA
35.38
USD
100201-964
100201-966
Calcium stearate ≥85%
Calcium stearate
Formula:
C₃₆H₇₀CaO₄ MW: 607.03 g/mol Melting Pt: 147 - 149 °C Density: 1.065 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00036390 CAS Number: 1592-23-0 EINECS: 216-472-8 Merck Index: 13,01708 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...