Calcium oxide ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Quicklime, Calcium oxide from marble, caustic, Lime
93-2031-1000G
93-2031-250G
100201-962EA
195.32
USD
100201-962
100204-226
Calcium oxide ≥98%
Calcium oxide
Formula:
CaO MW: 56.08 g/mol Boiling Pt: 2850 °C Melting Pt: 2580 °C Density: 3.38 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010911 CAS Number: 1305-78-8 EINECS: 215-138-9 UN: 1910 ADR: 8,III Merck Index: 13,01691 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...