Calcium iodide hydrate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

93-2025-250G 93-2025-50G
100198-328EA 444.43 USD
100198-328 100204-216
Calcium iodide hydrate ≥98%
Calcium iodide hydrate
Formula: CaI₂·H₂O
Density: 2.55 g/cm³ (20 °C)
MDL Number: MFCD00149622
CAS Number: 71626-98-7
EINECS: 233-276-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR