Calcium carbonate ≥99.95% (Ca basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-2011-100G
93-2011-500G
100209-040EA
96.25
USD
100209-040
100210-238
Calcium carbonate ≥99.95% (Ca basis)
Calcium carbonate
Formula:
CaCO₃ Melting Pt: 825 °C Density: 2.71 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010906 CAS Number: 471-34-1 EINECS: 207-439-9 Merck Index: 13,01658 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...