Potassium fluoride, anhydrous ≥99.97% (K basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-1982-25G 93-1982-5G
100210-232EA 478.4 USD
100210-232 100210-234
Potassium fluoride, anhydrous ≥99.97% (K basis)
Potassium fluoride
Formula: KF
MW: 58.1 g/mol
Boiling Pt: 1505 °C
Melting Pt: 846 °C
Density: 2.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011398
CAS Number: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III
Merck Index: 13,07714

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR