Potassium superoxide ≥96%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-1970-100G
93-1970-25G
93-1970-450G
100198-324EA
107.57
USD
100198-324
100197-728
100201-952
Potassium superoxide ≥96%
Potassium superoxide
Formula:
KO₂ Melting Pt: ~400 °C Density: 2.14 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00082411 CAS Number: 12030-88-5 EINECS: 234-746-5 UN: 2466 ADR: 5.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...