Potassium hexafluorosilicate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Potassium fluorosilicate

93-1959-500G 93-1959-2000G
100200-066EA 67.94 USD
100200-066 100210-226
Potassium hexafluorosilicate ≥99%
Potassium hexafluorosilicate
Formula: F₆K₂Si
MW: 220.27 g/mol
Melting Pt: Dec.
Density: 2.67 g/cm³ (20 °C)
MDL Number: MFCD00040666
CAS Number: 16871-90-2
EINECS: 240-896-2
UN: 2655
ADR: 6.1,III
Merck Index: 12,07803

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR